missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ E-A-R™ E-A-Rsoft Yellow Earplugs
Offers high attenuation and are made from a slow expanding, PU foam material. 3M E-A-R Polyurethane Earplug – 36dB.
Brand: 3M™ 7000038201
This item is not returnable.
View return policy
Description
- Made from a slow expanding, PU foam material
- Provides evenly distributed pressure, giving flexibility, and a good seal with optimum comfort.
- Bright, neon color makes it appealing to wearers

Specifications
Yellow Neon | |
250/Pk. | |
Uncorded | |
Polyurethane |
CE Approved | |
36 dB | |
Ear Plug | |
Roll-down |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ E-A-R™ E-A-Rsoft Yellow Earplugs > Color: ?Neon Yellow; Quantity: 250/Pk.
Spot an opportunity for improvement?Share a Content Correction