missing translation for 'onlineSavingsMsg'
Learn More

Zeiss™ Invertoskop™ Microscope Specimen Holders

Designed for a variety of applications

Brand:  Zeiss™ 471746-0000-000

 View more versions of this product

Product Code. 10544843

This item is not returnable. View return policy

Description

Description

Mounting frame M

471746

  • For microtiter plates
  • 96 positions
  • With adhesive scales

471718

  • Petri dishes dia. 24 to 68mm
  • Slides (max. length 120 mm)
Specifications

Specifications

Mounting Frame M
Invertoskop microscope
Mounting frame M, Adhesive scale
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers