missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 24 Deep-Well RB Block
Designed for experimental protocols that require volumes exceeding the capacity of conventional shallow well plates
Brand: Invitrogen™ CS15124
Includes: 25 Deep Well RB Blocks
Description
Compatible with magnetic bead-based technology provided in GeneCatcher™ gDNA 0.3-1mL Blood Kit and 24-well Magnetic Separator.
- Deep, round-bottom wells with a 10mL working volume per well
- Polypropylene plastic construction, conforming to the proposed SBS microplate standards
- Guaranteed compatibility with robotic handlers and centrifuge carriers
DNA and RNA Purification and Analysis, DNA Extraction, Genomic DNA Purification, gDNA from Blood (Medium-Scale, 100 μl-1mL)
Order Info
Shipping Condition: Room Temperature
Specifications
Contains 25 Deep Well RB Blocks. Store at room temperature. | |
Room Temperature | |
24 Deep Wells |
For Research Use Only. Not for use in diagnostic procedures.