missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gateway™ LR Clonase™ Enzyme mix

Enzyme mix that offers high recombination efficiency

Brand:  Invitrogen™ 11791043

1580.70 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10484202

  • £2395.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: Proteinase K solution (2μg/μL) and a positive control vector
This item is not returnable. View return policy

Description

Description

Gateway LR Clonase enzyme mix contains proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes that catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate your expression clone.

  • Specifically optimized for both single and multiple fragment cloning
  • Provided in optimized single mix of enzyme and reaction buffer, ensuring enzyme stability and ease of use with few pippetting steps for both single and multiple fragment cloning

Cloning, Gateway Cloning, Gateway Recombination

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

All Gateway™ LR Clonase™ enzyme kits include proteinase K solution (2 μg/μl) and a positive control vector. Store LR Clonase™ enzyme at -80°C; LR Clonase™ II or II Plus Enzyme mixtures at -20°C. Guaranteed stable for 6 months when properly stored.
LR Clonase
100 Reactions
Clonase™, Gateway™
Dry Ice
Reaction Buffer
LR Clonase Enzyme Mix
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.