missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gateway™ LR Clonase™ Enzyme mix
Enzyme mix that offers high recombination efficiency
£804.00 - £2395.00
Specifications
Content And Storage | All Gateway™ LR Clonase™ enzyme kits include proteinase K solution (2 μg/μl) and a positive control vector. Store LR Clonase™ enzyme at -80°C; LR Clonase™ II or II Plus Enzyme mixtures at -20°C. Guaranteed stable for 6 months when properly stored. |
---|---|
Shipping Condition | Dry Ice |
Enzyme | LR Clonase |
Compatible Buffer | Reaction Buffer |
Product Type | LR Clonase Enzyme Mix |
Includes: Proteinase K solution (2μg/μL) and a positive control vector
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10694223
|
Invitrogen™
11791019 |
20 Reactions |
£804.00
20 reactions |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10484202
|
Invitrogen™
11791043 |
100 Reactions |
£2395.00
100 reactions |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gateway LR Clonase enzyme mix contains proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes that catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate your expression clone.
- Specifically optimized for both single and multiple fragment cloning
- Provided in optimized single mix of enzyme and reaction buffer, ensuring enzyme stability and ease of use with few pippetting steps for both single and multiple fragment cloning
Cloning, Gateway Cloning, Gateway Recombination
Order Info
Shipping Conditions: Dry ice
Specifications
All Gateway™ LR Clonase™ enzyme kits include proteinase K solution (2 μg/μl) and a positive control vector. Store LR Clonase™ enzyme at -80°C; LR Clonase™ II or II Plus Enzyme mixtures at -20°C. Guaranteed stable for 6 months when properly stored. | |
LR Clonase | |
LR Clonase Enzyme Mix |
Dry Ice | |
Reaction Buffer | |
Clonase™, Gateway™ |
For Research Use Only. Not for use in diagnostic procedures.