Learn More
Invitrogen™ LR Clonase™ II Plus enzyme
Mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor), and Xis (Excisionase) enzymes.
Brand: Invitrogen™ 12538200
2065.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10000kg
Description
Gateway™ LR Clonase™ enzyme mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes that catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate your expression clone. We offer different formats of LR Clonase enzyme mixtures, depending on your application and desired format.
- Gateway™ LR Clonase™ enzyme mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes
- They catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate an expression clone
- Different formats of LR Clonase enzyme mixtures are offered depending on the application and desired format
- LR Clonase II Plus enzyme mix is the latest version that offers the highest recombination efficiency available
- It is specifically optimized for both single and multiple fragment cloning
- LR Clonase II Plus enzyme mix is provided in an optimized single mix of enzyme and reaction buffer
- This ensures enzyme stability and ease of use with few pippetting steps for both single and multiple fragment cloning
- Guaranteed stable for 6 months when properly stored at -20°C
Specifications
Includes LR Clonase II Plus enzyme and proteinase K solution (2 μg/μL). Store at -80°C. Guaranteed stable for 6 months when properly stored. | |
LR Clonase | |
100 Reactions | |
Clonase™, Gateway™ |
Dry Ice | |
Reaction Buffer | |
LR Clonase Enzyme |
For Research Use Only. Not for use in diagnostic procedures.