Learn More
Invitrogen™ LR Clonase™ II Plus enzyme
Mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor), and Xis (Excisionase) enzymes.
£562.00 - £2350.00
Specifications
Content And Storage | Includes LR Clonase II Plus enzyme and proteinase K solution (2 μg/μL). Store at -80°C. Guaranteed stable for 6 months when properly stored. |
---|---|
Shipping Condition | Dry Ice |
Enzyme | LR Clonase |
Compatible Buffer | Reaction Buffer |
Product Type | LR Clonase Enzyme |
Includes: Proteinase K solution (2μg/μL) and a positive control vector
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10108672
|
Invitrogen™
12538120 |
20 Reactions |
£562.00
20 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12176590
|
Invitrogen™
12538200 |
100 Reactions |
£2350.00
100 reactions |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gateway™ LR Clonase™ enzyme mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes that catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate your expression clone. We offer different formats of LR Clonase enzyme mixtures, depending on your application and desired format.
- Gateway™ LR Clonase™ enzyme mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes
- They catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate an expression clone
- Different formats of LR Clonase enzyme mixtures are offered depending on the application and desired format
- LR Clonase II Plus enzyme mix is the latest version that offers the highest recombination efficiency available
- It is specifically optimized for both single and multiple fragment cloning
- LR Clonase II Plus enzyme mix is provided in an optimized single mix of enzyme and reaction buffer
- This ensures enzyme stability and ease of use with few pippetting steps for both single and multiple fragment cloning
- Guaranteed stable for 6 months when properly stored at -20°C
Order Info
Shipping Conditions: Dry ice
Specifications
Includes LR Clonase II Plus enzyme and proteinase K solution (2 μg/μL). Store at -80°C. Guaranteed stable for 6 months when properly stored. | |
LR Clonase | |
LR Clonase Enzyme |
Dry Ice | |
Reaction Buffer | |
Clonase™, Gateway™ |
For Research Use Only. Not for use in diagnostic procedures.