missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG w/ROX

qPCR SuperMixes with ROX for ABI instruments are ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA.

Marke:  Invitrogen™ 11743100

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 11528656

  • £320.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Entdecken Sie weitere Sonderangebote

For Research Use Only. All usage must comply with product instructions.
 

Beinhaltet: Platinum™ Quantitative PCR SuperMix-UDG with ROX; Magnesium Chloride (50mM)
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

qPCR SuperMixes with ROX are available in two formats:
  • Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX provides a sensitive, cost-effective system for real-time PCR detection with SYBR™ Green I dye
  • Platinum™ qPCR SuperMix-UDG with ROX offers superior performance with either fluorogenic primers (e.g., LUX™ Primers) or probe-based detection systems (e.g., TaqMan™)

    Bestellinformation

    Guaranteed stable for 6 months when properly stored. Shipping Conditions: Dry Ice.

    TRUSTED_SUSTAINABILITY
Spezifikation

Spezifikation

• 2 × 1.25 ml Platinum™ Quantitative PCR SuperMix-UDG with ROX
• 1 ml Magnesium Chloride (50 mM)

Store at -20°C in the dark.
High
Platinum Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT System
Quantitative PCR SuperMix-UDG
100 Reactions
2X
100 Reactions
2.5 mL
Primer-probe
qPCR
Standard
Platinum™
Dry Ice
Gene Expression
2 X
DNA (Genomic), cDNA
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not for use in diagnostic procedures.