missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG w/ROX
qPCR SuperMixes with ROX for ABI instruments are ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA.
£320.00 - £1340.00
Specifications
Detection Method | Primer-probe |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Platinum Taq DNA Polymerase |
Reaction Speed | Standard |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11538656
|
Invitrogen™
11743500 |
500 Reactions |
£1340.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11528656
|
Invitrogen™
11743100 |
100 Reactions |
£320.00
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
qPCR SuperMixes with ROX are available in two formats:- Platinum™ SYBR™ Green qPCR SuperMix-UDG with ROX provides a sensitive, cost-effective system for real-time PCR detection with SYBR™ Green I dye
- Platinum™ qPCR SuperMix-UDG with ROX offers superior performance with either fluorogenic primers (e.g., LUX™ Primers) or probe-based detection systems (e.g., TaqMan™)
Order Info
Guaranteed stable for 6 months when properly stored. Shipping Conditions: Dry Ice.
Specifications
Primer-probe | |
qPCR | |
Standard | |
Platinum™ | |
Dry Ice | |
2X | |
DNA (Genomic), cDNA |
High | |
Platinum Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT System | |
Quantitative PCR SuperMix-UDG | |
Gene Expression | |
2 X |
For Research Use Only. Not for use in diagnostic procedures.