Learn More
Invitrogen™ IL-12 p40 Human ProQuantum Immunoassay Kit
Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument
Brand:  Invitrogen™ A355775
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 17153099
  • £2895.00 / Each
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
Interleukin-12 (IL-12) is a heterodimeric 70 kD (p70) cytokine composed of two covalently linked,glycosylated chains,40kD (p40) and 35-kD (p35). IL-12 is mainly produced by monocytes,macrophages,and dendritic cells in response to bacterial products such as lipopolysaccharides (LPS),to intracellular pathogens or upon interaction with activated T cells. IL-12 was originally discovered because of its ability to induce interferon-gamma production,cell proliferation,and cytotoxicity mediated by natural killer cells and T cells. It is now established that IL-12 also plays a key role in the development of Th1 responses,leading to IFN-gamma and IL-2 production. These cytokines can in turn promote T-cell responses and macrophage activation. The p40 and p35 subunits by themselves have no IL-12 bioactivity,though the p40 homodimer has been shown to bind the IL-12 receptor and to be an antagonist of IL-12 p70. Free p40 is typically secreted in vast excess of IL-12 p70 by cells co-expressing both the p35 and p40 subunits. The p40 subunit of IL-12 has been shown to have extensive amino acid sequence homology to the extracellular domain of the IL-6 receptor,while the p35 subunit shows some homology to IL-6 and G-CSF.
Specifications
Specifications
P29460
<0.05pg/mL
ProQuantum
Human
qPCR Instruments
CLMF; CLMF2; IL-12B; IMD28; IMD29; NKSF; NKSF2
FAM™
RUO
-20°C
30 min.
0.064 – 10000pg/mL
Taqman™ Real-time PCR
Plasma, Serum, Supernatant
Cytokine protein quantitation
3593
Assay Dilution Buffer<Br>Antibody-Conjugate A<Br>Antibody-Conjugate B<Br>Antibody-Conjugate Dilution Buffer<Br>Standard<Br>Master Mix<Br>Ligase
5 x 96 Tests
Serum, 2-5 μL; Supernatant, 2-5 μL; Plasma, 2-5 μL
Human
2 hr.