missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-12 p40 Human ProQuantum Immunoassay Kit
Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument
£648.00 - £2895.00
Specifications
Accession Number | P29460 |
---|---|
Assay Range | 0.064 – 10000pg/mL |
Assay Sensitivity | <0.05pg/mL |
Detection Method | Taqman™ Real-time PCR |
Product Type | ProQuantum |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980463
|
Invitrogen™
A35577 |
96 Tests |
£648.00
96 reactions |
In Stock
Log in to see stock. |
|||||
17153099
|
Invitrogen™
A355775 |
5 x 96 Tests |
£2895.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
Description
- High-sensitivity – detect low levels of proteins with greater sensitivity than traditional methods like ELISA
- Small sample consumption – use 2-5 μL of sample (e.g., 2 μL vs 75 μL for triplicate wells with other methods)
- Fast, easy workflow – no wash steps, 2 hours from sample to answer
- Broad dynamic range – 5 logs minimizes sample dilutions to ensure falling within the range
- No proprietary instrument to purchase – runs on any real-time PCR instrument
- Includes intuitive cloud-based software – for robust data analysis and statistical group wise comparison
Specifications
P29460 | |
<0.05pg/mL | |
ProQuantum | |
Human | |
qPCR Instruments | |
CLMF; CLMF2; IL-12B; IMD28; IMD29; NKSF; NKSF2 | |
FAM™ | |
Serum, 2-5 μL; Supernatant, 2-5 μL; Plasma, 2-5 μL | |
Human | |
2 hr. |
0.064 – 10000pg/mL | |
Taqman™ Real-time PCR | |
Plasma, Serum, Supernatant | |
Cytokine protein quantitation | |
3593 | |
Assay Dilution Buffer<Br>Antibody-Conjugate A<Br>Antibody-Conjugate B<Br>Antibody-Conjugate Dilution Buffer<Br>Standard<Br>Master Mix<Br>Ligase | |
RUO | |
-20°C | |
30 min. |