Learn More
Invitrogen™ PureLink™ Genomic DNA Mini Kit
The PureLink™ Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format.
Brand: Invitrogen™ K182000
31.75 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
The PureLink™ Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format. Features of the PureLink™ Genomic DNA Mini Kit include:
• Kit flexibility—one kit works with a variety of sample types and sizes
• Ultraclean gDNA—minimal contamination of DNA product means successful downstream application
• Improved design—optimized spin-column design and buffer formulation for better yield and purity
Multiple sample sources, one kit
The PureLink™ Genomic DNA Mini Kit can be used with many different sample types, each with its own optimized protocol outlined in the manual.
Optimized components and protocols
The PureLink™ Genomic DNA Mini Kit includes parts and protocols that improve upon the easy and familiar spin-column methods currently in use. Columns are newly configured to bind and release DNA more efficiently. Each sample source uses a specialized lysis procedure and an optimized lysis buffer formulated to enhance proteinase K activity and eliminate protein contamination. A chaotropic salt buffer promotes stable binding of DNA to the column resin, and powerful wash buffers remove all traces of protein and salt. DNA is eluted in a low-salt buffer to allow for pH stabilization of the DNA in storage.
Higher yields and purity
With PureLink™ Genomic DNA Kits you can expect high yields of high-purity gDNA (determined by A260/A280 measurements), no matter the sample type, from bacteria to tissues to blood to cultured cells. The enhanced PureLink™ Lysis and Wash Buffers help to consistently deliver purified gDNA samples with absorbance reading ratios of ∼1.9 for both A260/A280 and A260/A230, indicating no detectable contamination from carryover of protein or guanidine.
Order Info
Shipping Condition: Room Temperature

Specifications
• 2.0 mL PureLink™ Genomic Lysis/Binding Buffer • 1.8 mL PureLink™ Genomic Digestion Buffer • 2.0 mL PureLink™ Genomic Wash Buffer 1 • 1.5 mL PureLink™ Genomic Wash Buffer 2 • 2.0 mL PureLink™ Genomic Elution Buffer • 0.4 mL RNase A (20 mg/mL) • 0.2 mL Proteinase K (20 mg/mL) • 10 PureLink™ Spin Columns with Collection Tubes • 20 PureLink™ Collecion Tubes (2.0 mL) Store all components at room temperature. |
|
Mini | |
Genotyping, Real-Time Quantitative PCR (qPCR), Southern Blotting, Sequencing, PCR | |
100 μL to 1 mL Blood Samples (Medium-Scale) | |
Genomic DNA Mini Kit | |
Room Temperature |
Silica Spin Column | |
Bacteria, Blood, Cells, Tissue | |
Not High-throughput Compatible (Manual) | |
PureLink™ | |
10 Preps | |
Genomic DNA |
Safety and Handling
- Genomic DNA Mini Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Guanidine hydrochloride
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3113916 | Certificate of Analysis | 31/01/2025 | K182000 |
2926482 | Certificate of Analysis | 31/01/2025 | K182000 |
2270801A | Certificate of Analysis | 29/01/2025 | K182000 |
2270801A | Certificate of Analysis | 29/01/2025 | K182000 |
2365216 | Certificate of Analysis | 29/01/2025 | K182000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.