Learn More
Invitrogen™ SYBR™ Safe DNA Gel Stain in 0.5X TBE
Developed specifically for reduced mutagenicity – to be safer than ethidium bromide for staining DNA in agarose or acrylamide gels
Brand: Invitrogen™ S33100
93.10 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
SYBR Safe stain is less mutagenic than ethidium bromide and offers better sensitivity than ethidium bromide. Supplied as a premixed solution that can be used like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe stain has fluorescence excitation maxima at ∽280 and ∽502nm, and an emission maximum at ∽530nm. SYBR Safe DNA gel stain is also available as 4L (S-33101) of ready-to-use solution. The SYBR Safe DNA Gel Stain Starter Kit (S-33110, sold separately) includes 1L of SYBR Safe DNA gel stain and one photographic filter (S-37100).
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room temperature
Specifications
• Supplied as a ready-to-use solution in 0.5X TBE Store at room temperature in the original container. |
|
Fluorescence | |
DNA Gel Stain | |
1 L | |
DNA |
In-Gel Detection | |
SYBR Safe | |
Less hazardous | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.