missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Trypan Blue Stain (0.4%) for use with the Countess™ Automated Cell Counter

For use with the Countess™ Automated Cell Counter

Brand:  Invitrogen™ T10282

Code : 50

Additional Details : Weight : 0.50000kg


Product Code. 10702404

  • £24.22 / Each
In Stock
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

  • Used to identify dead cells
  • Detection Method: Colorimetric

Cell Counting, Viability and Cryopreservation

Specifications

Specifications

Stain
Blue
Not High-throughput Compatible (Manual)
Contents: 2 1-mL vials of 0.4% solution of Trypan blue stain. Room temperature storage.
Room Temperature
Countess™ Automated Cell Counter (C10227)
Colorimetric
Trypan Blue
2 x 1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Trypan Blue stain 0.4% *for use with Countess automated cell counter*

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 2, 7-Naphthalenedisulfonicacid, 3, 3-¢(3, 3-dimethyl¢1, 1-biphenyl!-4, 4-diyl)bis(azo)!bis¢5-amino-4

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.