Learn More
Invitrogen™ UltraPure™ Formamide
UltraPure Formamide is commonly used for denaturing nucleic acids for sequencing gel electrophoresis, electron microscopy, and hybridization.
Brand: Invitrogen™ 15515026
Description
UltraPure Formamide is commonly used for denaturing nucleic acids for sequencing gel electrophoresis, electron microscopy, and hybridization. A liquid at room temperature, UltraPure Formamide is vacuum distilled and packaged under dry nitrogen and can be used as supplied for many applications within three months of date of purchase. Because the breakdown products of formamide degrade nucleic acids, for sensitive applications formamide should be deionized by treatment with a mixed-bed ion-exchange resin immediately before use.
Order Info
Approved for shipment at Room Temperature or on Wet Ice

Specifications
Store in refrigerator (2–8°C). | |
Formamide | |
DNase-Free, RNase-Free | |
Approved for shipment at Room Temperature or on Wet Ice |
Cellular Imaging/Nucleic Acid Gel Electrophoresis & Blotting | |
UltraPure™ | |
500 g | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.