Learn More
Scientific Glass Labs™ Soda Lime Glass Watch Glasses
Manufactured from Soda lime glass. Scientific Glass Labs™ Soda Lime Glass Watch Glasses with thin, clear glass and ground edge can be used to evaporate liquids, hold solids for weighing, or for heating small substances.
Brand:  Scientific Glass Labs™ 170/1/200
 View more versions of this product
Product Code. 13152440
  • £419.00 / Pack of 5
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Watch glasses are useful for evaporating liquids, holding solids for weighing, or for heating small substances.
  • Manufactured from Soda lime glass
  • Thin, clear glass
  • Ground edge
Specifications
Specifications
Flat
Round
Evaporating Liquids, Heating Small Substances, Holding Solids for Weighing
200 mm
Soda Lime Glass
Watch Glass
Ground Edge
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Scientific Glass Labs™ Soda Lime Glass Watch Glasses > Diameter: 200mm