missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Glycerol

UltraPure Glycerol is suitable for the concentration and storage of enzymes at low temperatures. A 50% (v/v) solution will not freeze at -20°C.

Brand:  Invitrogen™ 15514029

244.00 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10779053

  • £305.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • UltraPure Glycerol is suitable for the concentration and storage of enzymes at low temperatures
  • A 50% (v/v) solution will not freeze at -20°C
  • Since it has a density of 1.26 g/mL, glycerol frequently is used as a component in electrophoresis loading buffers
  • UltraPure Glycerol gradients can be used to purify proteins, bacteriophage, or organelles
  • Redistilled UltraPure Glycerol is ≥ 99% pure
  • No DNase, RNase, or protease activity detected
  • Store at room temperature

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glycerol
UltraPure™
3.8 L
Liquid
Store at room temperature.
Molecular Biology Grade
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.