missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ SSPE, 20X

For use in nucleic acid hybridizations and blot transfer applications

Brand:  Invitrogen™ 15591027

 View more versions of this product

Product Code. 11578896

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • UltraPure 20X SSPE is prepared buffer concentrate
  • Supplied in 1L plastic bottles/4L/10L stackable Cubitainer™, Box
  • Sterile-filtered UltraPure 20X SSPE contains 3.0 M NaCl, 0.2M NaH2PO4, and 0.02M EDTA at pH 7.4
  • Performance and Quality Testing: No DNase or RNase activity detected. Sustainable packaging

DNA & RNA Purification & Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

20 X
Nucleic Acid Gel Electrophoresis, Blotting
10 L
Liquid
Store at room temperature.
UltraPure™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.