missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ SSPE, 20X
For use in nucleic acid hybridizations and blot transfer applications
£23.87 - £136.00
Specifications
Concentration | 20 X |
---|---|
Content And Storage | Store at room temperature. |
For Use With (Application) | Nucleic Acid Gel Electrophoresis, Blotting |
Product Line | UltraPure™ |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10256282
|
Invitrogen™
15591043 |
1 L |
£23.87
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11578896
|
Invitrogen™
15591027 |
10 L | N/A | ||||||
Description
- UltraPure 20X SSPE is prepared buffer concentrate
- Supplied in 1L plastic bottles/4L/10L stackable Cubitainer™, Box
- Sterile-filtered UltraPure 20X SSPE contains 3.0 M NaCl, 0.2M NaH2PO4, and 0.02M EDTA at pH 7.4
- Performance and Quality Testing: No DNase or RNase activity detected. Sustainable packaging
DNA & RNA Purification & Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
20 X | |
Nucleic Acid Gel Electrophoresis, Blotting | |
Room Temperature |
Store at room temperature. | |
UltraPure™ | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.