missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Urea

UltraPure™ Urea is an effective protein denaturant.

Brand:  Invitrogen™ 15505050

 View more versions of this product

Product Code. 17132571

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

UltraPure™ Urea is an effective protein denaturant. In a highly concentrated (6 to 8 M) urea solution, many proteins unfold and separate into their constituent polypeptide chains. UltraPure™ Urea is also used to denature nucleic acids in sequencing gels.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
UltraPure™
Room Temperature
Urea
2 kg
Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.