missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Urea

UltraPure™ Urea is an effective protein denaturant.

£48.81 - £138.64

Specifications

Product Type Urea
Product Line UltraPure™
Form Powder
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17132571
View Documents Promotion Details
Invitrogen™
15505050
2 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10469033
View Documents Promotion Details
Invitrogen™
15505035
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11548736
View Documents Promotion Details
Invitrogen™
15505027
2 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

UltraPure™ Urea is an effective protein denaturant. In a highly concentrated (6 to 8 M) urea solution, many proteins unfold and separate into their constituent polypeptide chains. UltraPure™ Urea is also used to denature nucleic acids in sequencing gels.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Urea
UltraPure™
Powder
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.