Learn More
Invitrogen™ X-Gal
X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays.
Brand: Invitrogen™ 15520018
180.88 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays. Bacteria containing active β-galactosidase produce blue colonies when grown on plates containing X-Gal. X-Gal is supplied as white crystals and is soluble in dimethylformamide.
Primary applications
- Blue-white screening—detect functional β-galactosidase (reporter gene expression)
- Histochemical detection of β-galactosidase activity in fixed tissues
- X-gal overlay assay
Find the media format that fits your needs
We offer many ready-to-use media formulations, such as LB Broth (10855001), Terrific Broth (A1374301) or S.O.C recovery medium (15544034).
We also offer IPTG (15529019) for blue-white screening.
Explore other media formats for bacterial growth.
Order Info
Approved for shipment at Room Temperature or on Wet Ice.
Specifications
5-bromo-4-chloro-3-indolyl-β-D-galactoside | |
Colorimetric | |
Bottle | |
X-Gal Substrate | |
Approved for shipment at Room Temperature or on Wet Ice |
• X-Gal, powder, 1 g | |
X-Gal, Cell-Permeant | |
Immunoblotting | |
1 g | |
Powder |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.