Learn More
Invitrogen™ X-Gal
X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays.
£61.50 - £238.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11538746
|
Invitrogen™
15520034 |
100 mg |
£61.50
100mg |
In Stock
Log in to see stock. |
|||||
10554973
|
Invitrogen™
15520018 |
1 g |
£238.00
1g |
In Stock
Log in to see stock. |
|||||
Description
X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays. Bacteria containing active β-galactosidase produce blue colonies when grown on plates containing X-Gal. X-Gal is supplied as white crystals and is soluble in dimethylformamide.
Primary applications
- Blue-white screening—detect functional β-galactosidase (reporter gene expression)
- Histochemical detection of β-galactosidase activity in fixed tissues
- X-gal overlay assay
Find the media format that fits your needs
We offer many ready-to-use media formulations, such as LB Broth (10855001), Terrific Broth (A1374301) or S.O.C recovery medium (15544034).
We also offer IPTG (15529019) for blue-white screening.
Explore other media formats for bacterial growth.
Order Info
Approved for shipment at Room Temperature or on Wet Ice.
Specifications
Colorimetric |