Learn More
Invitrogen™ X-Gal
X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays.
Brand:  Invitrogen™ 15520034
46.74 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 11538746
  • £61.50 / 100mg
In Stock
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays. Bacteria containing active β-galactosidase produce blue colonies when grown on plates containing X-Gal. X-Gal is supplied as white crystals and is soluble in dimethylformamide.
Primary applications
  • Blue-white screening—detect functional β-galactosidase (reporter gene expression)
  • Histochemical detection of β-galactosidase activity in fixed tissues
  • X-gal overlay assay
Find the media format that fits your needs
We offer many ready-to-use media formulations, such as LB Broth (10855001), Terrific Broth (A1374301) or S.O.C recovery medium (15544034).
We also offer IPTG (15529019) for blue-white screening.
Explore other media formats for bacterial growth.
Order Info
Approved for shipment at Room Temperature or on Wet Ice.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5-bromo-4-chloro-3-indolyl-β-D-galactoside
Colorimetric
Bottle
X-Gal Substrate
Approved for shipment at Room Temperature or on Wet Ice
Store in freezer (-5°C to -30°C).
X-Gal, Cell-Permeant
Immunocytochemical Assays