Learn More
Invitrogen™ Invitrolon™ PVDF/Filter Paper Sandwiches, 0.45 μm, 8.3 x 7.3 cm (for mini gels)
Pre-cut and pre-assembled and ready for protein blotting/transfer
Brand: Invitrogen™ LC2005
167.64 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00454kg
Includes: 20 membrane/filter paper sandwiches per pack
Description
Perform well for protein transferand particularly suitable for high-sensitivity and low-background immunoblotting.
Membranes are pre-cut to 8.3 cm x 7.3 cm to fit mini-gels and pre-assembled with two filter papers to simplify blotting setup. In addition, Invitrolon™ PVDF offers:
- Uniform 0.45 μm Pore Size—ideal for Western transfer of proteins >10 kDa
- High binding capacity—to enhance sensitivity and reduce background
- Reprobing characteristics—to eliminate repeat runs and transfers
- Durability and solvent-resistance—for protein sequencing and amino acid analysis
0.45 μm Pore Size. 8.3 x 7.3cm (for mini gels)
Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
8.3 cm | |
Sandwich | |
0.45 μm | |
Room Temperature | |
0.8 mm |
7.3 cm | |
PVDF | |
Invitrolon™ PVDF are conveniently supplied as 20 membrane/filter paper sandwiches per pack. Store at room temperature. | |
7.3 cm x 8.3 cm | |
Invitrolon Novex |
For Research Use Only. Not for use in diagnostic procedures.