missing translation for 'onlineSavingsMsg'
Learn More
Iodide Iodate concentrate, For 1L standard solution, 0.05 M I2 (0.1N), Honeywell Fluka™ Product Code.: 15605210
compliance-icons compliance-icons

Iodide Iodate concentrate, For 1L standard solution, 0.05 M I2 (0.1N), Honeywell Fluka™

Product Code. 15605210 Shop All Honeywell Fluka Products
6 x 1 Ea.
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
This item is not returnable. View return policy

Product Code. 15605210

Brand: Honeywell Fluka 380616X1EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for 1L standard solution, 0.05 M I2 (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0, 7553-56-2, 7758-05-6, 7732-18-5
Molecular Formula IK
MDL Number MFCD00011355
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
SMILES [K+].[I-]

Specifications

CAS Min % >10.0000%
CAS Max % <30.0000%, <20.0000%
Physical Form Concentrate
Quantity 6 x 1 Ea.
UN Number NONH for all modes of transport
Beilstein 3587194
IUPAC Name potassium;iodide
Molecular Weight (g/mol) 166.003
Packaging Ampoule
Chemical Name or Material Iodide Iodate concentrate, For 1L standard solution, 0.05 M I2 (0.1N)
compliance-icons compliance-icons
Product Identifier
  • Iodide Iodate concentrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Potassium iodate, Potassium iodide, iodine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodide Iodate concentrate, For 1L standard solution, 0.05 M I2 (0.1N), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.