missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 99.5+%, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10694722
72.06 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
100 g
250 g
500 g
7553-56-2
I2
253.81
MFCD00011355 MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
807
CHEBI:17606
II
7553-56-2
I2
253.81
MFCD00011355 MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
807
CHEBI:17606
II
Boiling Point 185°C
Melting Point 113°C
Color Gray
pH 5.1
Physical Form Solid
Quantity 100 g
IUPAC Name molecular iodine
Formula Weight 253.81g/mol
Packaging Amber glass bottle
Vapor Pressure 0.41 hPa 25
Density Vapor Density: 8.8
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Iodine
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'