Learn More
Iodine monobromide, 98%, Thermo Scientific Chemicals
Description
Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.
Solubility
Soluble in water, alcohol, ether, carbon disulfide and glacial acetic acid.
Notes
Air, light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, phosphorus, sodium and sodiumoxides and potassium.

Specifications
Specifications
Boiling Point | 116°C (decomposition) |
Melting Point | 50°C (sublimation) |
Quantity | 25 g |
UN Number | UN3260 |
Beilstein | 3902973 |
Flash Point | None |
Merck Index | 14,5016 |
Solubility Information | Soluble in water,alcohol,ether,carbon disulfide and glacial acetic acid. |
Formula Weight | 206.81 |
Percent Purity | 98% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10249542 | Certificate of Analysis | 05/03/2024 | A16158.30,A16158.22,A16158.14 |
10246901 | Certificate of Analysis | 30/06/2023 | A16158.30,A16158,A16158.22,A16158.14 |
10233159 | Certificate of Analysis | 06/02/2023 | A16158.30,A16158.22,A16158.14 |
10244815 | Certificate of Analysis | 28/11/2022 | A16158.30,A16158.22,A16158.14 |
M14I001 | Certificate of Analysis | 12/04/2022 | A16158.30,A16158.22,A16158.14 |
Safety and Handling


- Iodine monobromide
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Iodine bromide (IBr)
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.