missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Monochloride, Reagent grade, ≥95%, Honeywell™
£20.82 - £105.86
Chemical Identifiers
CAS | 7790-99-0 |
---|---|
Molecular Formula | ClI |
Molecular Weight (g/mol) | 162.35 |
MDL Number | MFCD00011354 |
InChI Key | QZRGKCOWNLSUDK-UHFFFAOYSA-N |
Synonym | iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french |
PubChem CID | 24640 |
IUPAC Name | iodochlorane |
SMILES | ClI |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15661830
|
Honeywell Research Chemicals
208221-5G |
5 g | Glass Bottle | ||||||
15641830
|
Honeywell Research Chemicals
208221-100G |
100 g | Glass Bottle | ||||||
15651830
|
Honeywell Research Chemicals
208221-250G |
250 g | Glass Bottle | ||||||
Description
Chemical Identifiers
7790-99-0 | |
162.35 | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
24640 | |
ClI |
ClI | |
MFCD00011354 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
iodochlorane |
Specifications
>97°C | |
7790-99-0 | |
Liquid or Solid | |
ICl | |
UN1792 | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
iodochlorane | |
24640 | |
≥95% | |
3.24g/mL at 25°C |
27°C | |
Brown to Red | |
ClI | |
MFCD00011354 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
ClI | |
162.35 | |
162.36g/mol | |
Reagent | |
Iodine Monochloride |
Safety and Handling
P261-P280-P305 + P351 + P338-P310
H314-H334
missing translation for 'dotInformation' : 8/PGII
missing translation for 'einecsNumber' : 232-236-7
missing translation for 'rtecsNumber' : NN1650000