missing translation for 'onlineSavingsMsg'
Learn More
Iodine monochloride, Thermo Scientific Chemicals Product Code.: 11383478

Iodine monochloride, Thermo Scientific Chemicals

Product Code. 11383478
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11383478

Brand: Thermo Scientific Alfa Aesar 039104.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ICl is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode frenchShow MoreShow Less
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Boiling Point 96°C to 98°C
Melting Point 25°C to 27°C
Physical Form Crystalline, Liquid
Quantity 500 g
Beilstein 3902972
Solubility Information Soluble in water,alcohol,ether,carbon disulfide.
Formula Weight 162.36
Sensitivity Light and moisture sensitive
Density 3.1822 g/mL
Chemical Name or Material Iodine monochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS27K040 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number039104.22,039104.14,039104.36
Lot NumberY01J013 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog Number039104.22,039104.14,039104.36
Lot NumberW12J003 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number039104.22,039104.14,039104.36
Lot NumberW12J002 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number039104.22,039104.14,039104.36
Lot Number10246980 Certificate TypeCertificate of Analysis Date16/06/2023 Catalog Number039104.22,039104.14,039104.36,039104

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodine monochloride, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.