missing translation for 'onlineSavingsMsg'
Learn More
Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals Product Code.: 11437097

Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11437097
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11437097

Brand: Thermo Scientific Alfa Aesar A14444.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and magnesium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-03-6
Molecular Formula C2H5I
Molecular Weight (g/mol) 155.966
MDL Number MFCD00001091
InChI Key HVTICUPFWKNHNG-UHFFFAOYSA-N
Synonym ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czechShow MoreShow Less
PubChem CID 6340
ChEBI CHEBI:42487
IUPAC Name iodoethane
SMILES CCI

Specifications

Melting Point -108°C
Density 1.95
Boiling Point 70°C to 73°C
Flash Point >73°C (163°F)
Refractive Index 1.512
Quantity 100 g
Beilstein 505934
Sensitivity Light and moisture sensitive
Merck Index 14,3813
Solubility Information Miscible with water.
Formula Weight 155.97
Percent Purity ≥98%
Chemical Name or Material Iodoethane, stabilized with copper
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242066 Certificate TypeCertificate of Analysis Date15/07/2022 Catalog NumberA14444.22,A14444.0E,A14444.36
Lot NumberB26Z033 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14444.22,A14444.0E,A14444.36
Lot NumberD2980C Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14444.22,A14444.0E,A14444.36
Lot NumberL23Z042 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14444.22,A14444.0E,A14444.36
Lot NumberUSLY001269 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14444.22,A14444.0E,A14444.36
compliance-icons compliance-icons
Product Identifier
  • Iodoethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Iodoethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.