missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane-d3, for NMR, 99+ atom % D, Thermo Scientific Chemicals

£57.49 - £171.86

Chemical Identifiers

CAS 865-50-9
Molecular Formula CH3I
Molecular Weight (g/mol) 144.96
MDL Number MFCD00001074
InChI Key INQOMBQAUSQDDS-FIBGUPNXSA-N
Synonym iodomethane-d3, iodo 2h3 methane, trideuterio iodo methane, iodo 2 h? methane, methyl-d3 iodide, trideuteromethyl iodide, iodomethane-12c,d3, methyl-12c,d3 iodide, iodoethane-d5 ethyliodid-d5, methyl iodide d3 stored over copper
PubChem CID 2723978
IUPAC Name trideuterio(iodo)methane
SMILES [2H]C([2H])([2H])I
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10780871
View Documents
Thermo Scientific Acros
174950050
5 g Glass Bottle
£57.49
5g
Please to purchase this item. Need a web account? Register with us today!
10790871
View Documents
Thermo Scientific Acros
174950250
25 g Glass Bottle
£171.86
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
Specifications

Chemical Identifiers

865-50-9
144.96
INQOMBQAUSQDDS-FIBGUPNXSA-N
2723978
[2H]C([2H])([2H])I
CH3I
MFCD00001074
iodomethane-d3, iodo 2h3 methane, trideuterio iodo methane, iodo 2 h? methane, methyl-d3 iodide, trideuteromethyl iodide, iodomethane-12c,d3, methyl-12c,d3 iodide, iodoethane-d5 ethyliodid-d5, methyl iodide d3 stored over copper
trideuterio(iodo)methane

Specifications

Liquid
2.3290g/mL
1.5262
865-50-9
CH3I
MFCD00001074
INQOMBQAUSQDDS-FIBGUPNXSA-N
trideuterio(iodo)methane
2723978
NMR
Colorless
99.0% min.
0.05% max. (Coulometric)
42.0°C
CD3I
iodomethane-d3, iodo 2h3 methane, trideuterio iodo methane, iodo 2 h? methane, methyl-d3 iodide, trideuteromethyl iodide, iodomethane-12c,d3, methyl-12c,d3 iodide, iodoethane-d5 ethyliodid-d5, methyl iodide d3 stored over copper
[2H]C([2H])([2H])I
144.96
144.96
Iodomethane-d3, 99+ atom % D
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Fatal if inhaled.
Harmful in contact with skin.
Suspected of causing cancer.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 212-744-5

Product Identifier
  • Iodomethane-d3

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only