Learn More
Iodotrimethylplatinum(IV), Thermo Scientific Chemicals
Description
They are used as catalysts in organic reactions. It is used in chemical, organic Intermediates research. They are used as platinum group compounds for the preparation of materials suitable for industrial applications, mainly in electronics as coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Orange to White |
Physical Form | Crystalline |
Quantity | 1 g |
Linear Formula | (CH3)3PtI |
Solubility Information | It is insoluble in water.It is sparingly soluble ether,alcohol,acetone or light petroleum. |
Formula Weight | 367.09 |
Odor | Odorless |
Sensitivity | Air & Light Sensitive |
Chemical Name or Material | Iodotrimethylplatinum(IV) |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10177329 | Certificate of Analysis | 08/12/2021 | 039286.02,039286.03,039288.MD |
G17S002 | Certificate of Analysis | 07/12/2021 | 039286.02,039286.03,039288.MD |
M22B036 | Certificate of Analysis | 29/11/2021 | 039286.02,039286.03,039288.MD |
L27P08 | Certificate of Analysis | 24/11/2021 | 039286.02,039286.03,039288.MD |
J11S004 | Certificate of Analysis | 23/11/2021 | 039286.02,039286.03,039288.MD |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.