missing translation for 'onlineSavingsMsg'
Learn More
Iodotrimethylplatinum(IV), Thermo Scientific Chemicals Product Code.: 11393688

Iodotrimethylplatinum(IV), Thermo Scientific Chemicals

Product Code. 11393688
0.5 g
Click to view available options
Quantity:
0.5 g
1 g
This item is not returnable. View return policy

Product Code. 11393688

Brand: Thermo Scientific Alfa Aesar 039286.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

They are used as catalysts in organic reactions. It is used in chemical, organic Intermediates research. They are used as platinum group compounds for the preparation of materials suitable for industrial applications, mainly in electronics as coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14364-93-3
Molecular Formula C3H9IPt
Molecular Weight (g/mol) 367.10
MDL Number MFCD00013702
InChI Key ZCSQPOLLUOLHHF-UHFFFAOYSA-M
Synonym iodotrimethylplatinum iv, iodotrimethylplatinum, trimethylplatinum iodide, acmc-1c6hk, trimethylplatinum iv iodide
PubChem CID 11824720
IUPAC Name carbanide;iodoplatinum
SMILES [CH3-].[CH3-].[CH3-].I[Pt]

Specifications

Color Orange to White
Physical Form Crystalline
Quantity 0.5 g
Linear Formula (CH3)3PtI
Solubility Information It is insoluble in water.It is sparingly soluble ether,alcohol,acetone or light petroleum.
Formula Weight 367.09
Odor Odorless
Sensitivity Air & Light Sensitive
Chemical Name or Material Iodotrimethylplatinum(IV)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10177329 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039286.02,039286.03,039288.MD
Lot NumberG17S002 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039286.02,039286.03,039288.MD
Lot NumberM22B036 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number039286.02,039286.03,039288.MD
Lot NumberL27P08 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number039286.02,039286.03,039288.MD
Lot NumberJ11S004 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number039286.02,039286.03,039288.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodotrimethylplatinum(IV), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.