missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva IPG Buffers
Produce more uniform conductivity along the Immobiline™ DryStrip during focusing. Cytiva IPG Buffers are ampholyte-containing buffers formulated for use with Immobiline™ DryStrip gels.
Brand: Cytiva 17-6000-88
This item is not returnable.
View return policy
Description
- Eliminates background staining
- Use with Immobiline DryStrip gels to improve protein solubility
- Ensures optimal conditions for IEF separation
- High sample loading capacity, improved resolution, and increases the probability of detecting low abundance proteins when used with Immobiline DryStrip gels
Order Info
Supplied in 1mL aliquots of water; order Immobiline Drystrip gels separately.
Specifications
Colorless to Light Yellow | |
3 to 10 nonlinear | |
Liquid | |
1mL |
Immobiline DryStrip Gels | |
IPG Buffer | |
IEF, 2-D Electrophoresis | |
2° to 8°C |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.