missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva IPG Buffers
Produce more uniform conductivity along the Immobiline™ DryStrip during focusing. Cytiva IPG Buffers are ampholyte-containing buffers formulated for use with Immobiline™ DryStrip gels.
£59.09
Specifications
Color | Colorless to Light Yellow |
---|---|
For Use With (Equipment) | Immobiline DryStrip Gels |
Product Type | IPG Buffer |
Form | Liquid |
For Use With (Application) | IEF, 2-D Electrophoresis |
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
10554565
|
Cytiva
17-6001-78 |
6 to 11 |
£59.09
1mL |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10534565
|
Cytiva
17-6000-86 |
4 to 7 |
£59.09
1mL |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10084334
|
Cytiva
17-6000-87 |
3 to 10 |
£59.09
1mL |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10544565
|
Cytiva
17-6000-88 |
3 to 10 nonlinear |
£59.09
Each |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10014104
|
Cytiva
17-6004-40 |
3 to 11 nonlinear |
£59.09
1mL |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Eliminates background staining
- Use with Immobiline DryStrip gels to improve protein solubility
- Ensures optimal conditions for IEF separation
- High sample loading capacity, improved resolution, and increases the probability of detecting low abundance proteins when used with Immobiline DryStrip gels
Order Info
Supplied in 1mL aliquots of water; order Immobiline Drystrip gels separately.
Specifications
Colorless to Light Yellow | |
IPG Buffer | |
IEF, 2-D Electrophoresis | |
2° to 8°C |
Immobiline DryStrip Gels | |
Liquid | |
1mL |