Learn More
Iridium black, 99.8% (metals basis), Thermo Scientific Chemicals
Description
The primary use of iridium is as a hardening agent in platinum alloys. Other uses are for the making of crucibles and devices that require high temperatures. Osmium/iridium alloys are used for compass bearings. Iridium is commonly used in complexes like Ir(mppy)3 and other complexes in polymer LED technology to increase the efficiency from 25% to about 100% due to triplet harvesting. Used in high-dose radiation therapy for the treatment of prostate and other forms of cancer. Iridium is used in tips of ballpoint pens. Iridium is used as a catalyst for carbonylation of methanol to create acetic acid. At one time, iridium as an alloy with platinum, was used in bushing the vents of heavy ordnance and, in a finely powdered condition (iridium black) for painting porcelain black.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 2 g |
UN Number | UN3089 |
Solubility Information | Soluble in aqua regia. |
Percent Purity | 99.8% |
Assay | (metals basis) |
Chemical Name or Material | Iridium black |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M16K025 | Certificate of Analysis | 11/06/2024 | 012071.09,012071.04,012071.02 |
W23I033 | Certificate of Analysis | 13/10/2022 | 012071.09,012071.04,012075.03,012071.02 |
W23G032 | Certificate of Analysis | 12/04/2022 | 012071.09,012071.04,012075.03,012071.02 |
I19M16 | Certificate of Analysis | 28/12/2021 | 012071.09,012071.04,012075.03,012071.02 |
61200905 | Certificate of Analysis | 28/12/2021 | 012071.09,012071.04,012075.03,012071.02 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.