missing translation for 'onlineSavingsMsg'
Learn More
Iridium(III) 2,4-pentanedionate, Ir 37.5% min, Thermo Scientific Chemicals Product Code.: 11396018

Iridium(III) 2,4-pentanedionate, Ir 37.5% min, Thermo Scientific Chemicals

Product Code. 11396018
1 g
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy

Product Code. 11396018

Brand: Thermo Scientific Alfa Aesar 010514.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a photoreducing agent, a precursor for preparing Ir-C thin film and a catalyst for hydrogenation of sulfur containing compounds. Used in various catalysis reactions in organic synthesis, including carbon nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15635-87-7
Molecular Formula C15H21IrO6
Molecular Weight (g/mol) 489.54
MDL Number MFCD00015353
InChI Key HLYTZTFNIRBLNA-LNTINUHCSA-K
Synonym Iridium(III) acetylacetonate
PubChem CID 117590592
SMILES [Ir+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 269°C to 271°C
Color Orange-Yellow
Physical Form Crystalline
Quantity 1 g
Solubility Information Insoluble in water; soluble in chloroform and benzene; slightly soluble in ethanol and methanol.
IUPAC Name iridium;(Z)-4-oxopent-2-en-2-olate
Formula Weight 489.53
Concentration or Composition (by Analyte or Components) Ir 37.5% min.
Odor Odorless
Chemical Name or Material Iridium(III) 2,4-pentanedionate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ28J008 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog Number010514.01,010514.03,010514.06
Lot NumberM28I064 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010516.14,010514.01,010514.03,010514.06
Lot NumberK09Y019 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010516.14,010514.01,010514.03,010514.06
Lot NumberB04X005 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010516.14,010514.01,010514.03,010514.06
Lot NumberT17F042 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010516.14,010514.01,010514.03,010514.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iridium(III) 2,4-pentanedionate, Ir 37.5% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.