missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) chloride tetrahydrate, 99+%, Thermo Scientific Chemicals

£26.43 - £425.12

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2Fe·4H2O
Molecular Weight (g/mol) 198.81
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
Synonym ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
PubChem CID 16211588
ChEBI CHEBI:86249
IUPAC Name dichloroiron;tetrahydrate
SMILES O.O.O.O.Cl[Fe]Cl
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10348150
View Documents
Thermo Scientific Acros
205080050
5 g Glass bottle
£26.43
5g
Please to purchase this item. Need a web account? Register with us today!
10401251
View Documents Promotion Details
Thermo Scientific Acros
205082500
250 g Glass bottle
£40.90
250g
Please to purchase this item. Need a web account? Register with us today!
10338150
View Documents
Thermo Scientific Acros
205080010
1 kg Glass bottle
£119.60
1kg
Please to purchase this item. Need a web account? Register with us today!
11994181
View Documents
Thermo Scientific Acros
205080051
5 kg Plastic bottle
£425.12
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

13478-10-9
198.81
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
CHEBI:86249
O.O.O.O.Cl[Fe]Cl
Cl2Fe·4H2O
WSSMOXHYUFMBLS-UHFFFAOYSA-L
16211588
dichloroiron;tetrahydrate

Specifications

105.0°C to 110.0°C
Yellow to Green
99+%
FeCl2·4H2O
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
WSSMOXHYUFMBLS-UHFFFAOYSA-L
dichloroiron;tetrahydrate
16211588
198.81
Iron(II) chloride tetrahydrate
13478-10-9
Crystalline Powder, Crystals and/or Chunks
Cl2Fe·4H2O
14,4043
Solubility in water: 1600g/L (10°C) 4100g/L (100°C). Other solubilities: soluble in alcohol
O.O.O.O.Cl[Fe]Cl
198.81
CHEBI:86249
≥99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

Product Identifier
  • Iron(II) chloride tetrahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.