Learn More
Iron(II) sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 64.0°C |
Color | Blue to Green |
Physical Form | Crystals |
Quantity | 250 g |
Assay Percent Range | 99.5% |
Linear Formula | FeSO4·7H2O |
Fieser | 01,393; 04,237; 05,308 |
Merck Index | 15,4084 |
Solubility Information | Solubility in water: 25.6 g/100mL (20°C). Other solubilities: insoluble in alcohol |
IUPAC Name | λ²-iron(2+) heptahydrate sulfate |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0469386 | Certificate of Analysis | 31/01/2025 | 201390010,201392500,201390051 |
A0461877 | Certificate of Analysis | 15/04/2024 | 201390010,201392500,201390051 |
A0459298 | Certificate of Analysis | 19/01/2024 | 201390010,201392500,201390050,201390051 |
A0457042 | Certificate of Analysis | 09/11/2023 | 201390010,201392500,201390050,201390051 |
A0453050 | Certificate of Analysis | 11/05/2023 | 201390010,201392500,201390051 |
Safety and Handling

- Iron(II) sulfate heptahydrate
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.