missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

£11.60 - £133.67

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10752051
View Documents Promotion Details
Thermo Scientific Acros
201390050
5 g Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10782241
View Documents Promotion Details
Thermo Scientific Acros
201392500
250 g Plastic bottle
£25.02
250g
Please to purchase this item. Need a web account? Register with us today!
10286300
View Documents Promotion Details
Thermo Scientific Acros
201390010
1 kg Plastic bottle
£39.40
1kg
Please to purchase this item. Need a web account? Register with us today!
11994241
View Documents Promotion Details
Thermo Scientific Acros
201390051
5 kg Plastic bottle
£133.67
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-63-0
278.01
SURQXAFEQWPFPV-UHFFFAOYSA-L
62662
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
FeH14O11S
MFCD00149719
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
CHEBI:75836

Specifications

7782-63-0
Crystals
FeH14O11S
MFCD00149719
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
278.01
CHEBI:75836
≥99.5%
Iron(II) sulfate heptahydrate
Blue to Green
99.5%
FeSO4·7H2O
01,393; 04,237; 05,308
SURQXAFEQWPFPV-UHFFFAOYSA-L
λ²-iron(2+) heptahydrate sulfate
62662
278.01
Analytical
Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only