missing translation for 'onlineSavingsMsg'
Learn More
Iron(II) sulfide, tech., Thermo Scientific Chemicals Product Code.: 11443037

Iron(II) sulfide, tech., Thermo Scientific Chemicals

Product Code. 11443037
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11443037

Brand: Thermo Scientific Alfa Aesar A15569.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.

Solubility
Soluble in water(0.0062g/L ), in acids with evolution of hydrogen sulfide. Insoluble in nitric acid.

Notes
Moisture Sensitive. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agent, acids, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-37-9
Molecular Formula FeS
Molecular Weight (g/mol) 87.91
MDL Number MFCD00011013
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfideShow MoreShow Less
PubChem CID 14828
SMILES [S--].[Fe++]

Specifications

Melting Point 1195°C
Quantity 250 g
Merck Index 14,4058
Solubility Information Soluble in water(0.0062g/L ),in acids with evolution of hydrogen sulfide. Insoluble in nitric acid.
IUPAC Name λ²-iron(2+) sulfanediide
Formula Weight 87.91
Grade Technical
Odor Odorless
Sensitivity Air and moisture sensitive
Density 4.74
Chemical Name or Material Iron(II) sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253428 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA15569.30,A15569.0B,A15569.0I
Lot Number10242481 Certificate TypeCertificate of Analysis Date28/07/2022 Catalog NumberA15569.30,A15569.0B,A15569.0I
Lot Number10237566 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15569.30,A15569.0B,A15569.0I
Lot NumberA2846A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15569.30,A15569.0B,A15569.0I
Lot Number10217497 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15569.30,A15569.0B,A15569.0I

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(II) sulfide, tech., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.