missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) chloride hexahydrate, 97+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid

Specifications
Specifications
Boiling Point | 280.0°C to 285.0°C |
Melting Point | 37.0°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 5 g |
Assay Percent Range | 97.0 to 102% (Iodometry) |
Linear Formula | FeCl3·6H2O |
Fieser | 01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158 |
Merck Index | 15,4046 |
Solubility Information | Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0454972 | Certificate of Analysis | 31/01/2025 | 423700025,423700100,423700050,423705000 |
A0463051 | Certificate of Analysis | 23/05/2024 | 423700025,423700100,423700050,423705000 |
A0454972 | Certificate of Analysis | 25/08/2023 | 423700025,423700100,423700050,423705000 |
A0438748 | Certificate of Analysis | 25/08/2023 | 423700025,423700100,423700050,423705000 |
A0452809 | Certificate of Analysis | 05/05/2023 | 423700025,423700100,423700050,423705000 |
Safety and Handling


Product Identifier
- Iron(III) chloride hexahydrate
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Iron(III) chloride hexahydrate, 97+%, ACS reagent, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction