missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals

£22.41 - £194.97

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10695862
View Documents
Thermo Scientific Acros
217091000
100 g Plastic bottle
£22.41
100g
Please to purchase this item. Need a web account? Register with us today!
10115180
View Documents
Thermo Scientific Acros
217095000
500 g Plastic bottle
£63.11
500g
Please to purchase this item. Need a web account? Register with us today!
10434605
View Documents
Thermo Scientific Acros
217090025
2.5 kg Plastic bottle
£194.97
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid for analysis
Specifications

Chemical Identifiers

10025-77-1
270.29
NQXWGWZJXJUMQB-UHFFFAOYSA-K
iron(3+) hexahydrate trichloride
Cl3FeH12O6
MFCD00149712
Ferric chloride hexahydrate
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

280.0°C to 285.0°C
10025-77-1
Adhering Crystalline Powder and Lumps
Cl3FeH12O6
MFCD00149712
15, 4046
Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
270.29
≥99%
Iron(III) chloride hexahydrate, p.a.
37.0°C
Orange
99.0 to 102% (Iodometry)
FeCl3·6H2O
01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158
Ferric chloride hexahydrate
NQXWGWZJXJUMQB-UHFFFAOYSA-K
iron(3+) hexahydrate trichloride
270.29
Analytical
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa

GHS Signal Word: Danger

missing translation for 'rtecsNumber' : NO5425000

Product Identifier
  • Iron(III) chloride hexahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Iron (III) chloridehexahydrate, Iron(III) chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only