missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) isopropoxide, 2.5% w/v in isopropanol, Thermo Scientific Chemicals

£30.05 - £99.90

Chemical Identifiers

CAS 14995-22-3
Molecular Formula C9H21FeO3
Molecular Weight (g/mol) 233.109
MDL Number MFCD00070436
InChI Key QUHDSMAREWXWFM-UHFFFAOYSA-N
Synonym iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe
PubChem CID 11961763
IUPAC Name iron(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11344668
View Documents
Thermo Scientific Alfa Aesar
043024.AA
5 mL
£30.05
5mL
Please to purchase this item. Need a web account? Register with us today!
11354668
View Documents
Thermo Scientific Alfa Aesar
043024.AC
25 mL
£99.90
25mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iron(III) isopropoxide is used in nano-materials & thin films, engineering & environmental, catalysis & synthesis and energy conversion & storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Chemical Identifiers

14995-22-3
233.109
QUHDSMAREWXWFM-UHFFFAOYSA-N
11961763
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3]
C9H21FeO3
MFCD00070436
iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe
iron(3+);propan-2-olate

Specifications

14995-22-3
0.79 g/mL
12°C (54°F)
C9H21FeO3
UN1219
iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe
QUHDSMAREWXWFM-UHFFFAOYSA-N
iron(3+);propan-2-olate
11961763
2.5% w/v in isopropanol
Iron (III) isopropoxide
Red to Brown
82°C
Alcohol-like
MFCD00070436
Moisture sensitive
It hydrolyzes with water.
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3]
233.109
233.11
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H225-H319-H335-H336

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

Product Identifier
  • Iron(III) isopropoxide, 2.5% w/v in isopropanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : 2-Propanol, Iron(III) isopropoxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only