Learn More
Iron(III) nitrate nonahydrate, 99+%, for analysis, Thermo Scientific Chemicals
£36.20 - £399.00
Chemical Identifiers
CAS | 7782-61-8 |
---|---|
Molecular Formula | FeH18N3O18 |
Molecular Weight (g/mol) | 403.99 |
MDL Number | MFCD00149708 |
InChI Key | SZQUEWJRBJDHSM-UHFFFAOYSA-N |
Synonym | ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate |
PubChem CID | 16211566 |
SMILES | O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10154170
|
Thermo Scientific Acros
202492500 |
250 g | Plastic bottle |
£36.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10256400
|
Thermo Scientific Acros
202490010 |
1 kg | Plastic bottle |
£108.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11934271
|
Thermo Scientific Acros
202490050 |
5 kg | Plastic bottle |
£399.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-61-8 | |
403.99 | |
SZQUEWJRBJDHSM-UHFFFAOYSA-N | |
16211566 |
FeH18N3O18 | |
MFCD00149708 | |
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate | |
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
47.0°C | |
Purple | |
99% min. (Iodometry) | |
Fe(NO3)3·9H2O | |
15,4054 | |
Solubility in water: soluble. Other solubilities: very soluble in ethanol, acetone | |
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
403.99 | |
404 | |
Analytical | |
1.6800g/mL |
7782-61-8 | |
Slightly Adhering Crystalline Powder | |
FeH18N3O18 | |
MFCD00149708 | |
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate | |
SZQUEWJRBJDHSM-UHFFFAOYSA-N | |
iron(3+) nonahydrate trinitrate | |
16211566 | |
≥99% | |
1.68 | |
Iron(III) nitrate nonahydrate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May intensify fire; oxidizer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cauti
GHS Signal Word: Warning
- Iron(III) nitrate nonahydrate
Signal Word
- Warning
Hazard Category
- Oxidising solid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Iron(III) nitrate nonahydrate, Ferric nitrate
RUO – Research Use Only