missing translation for 'onlineSavingsMsg'
Learn More
Iron(III) sulfate, Puratronic™, 99.998% (metals basis), Thermo Scientific™ Product Code.: 11390989

Iron(III) sulfate, Puratronic™, 99.998% (metals basis), Thermo Scientific™

Product Code. 11390989
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11390989

Brand: Thermo Scientific Alfa Aesar 010717.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-22-5
Molecular Formula Fe2O12S3
Molecular Weight (g/mol) 399.858
MDL Number MFCD00011007
InChI Key RUTXIHLAWFEWGM-UHFFFAOYSA-H
Synonym ferric sulfate, iron iii sulfate, iron persulfate, iron tersulfate, diiron tris sulphate, diiron trisulfate, ferric persulfate, ferric tersulfate, iron sesquisulfate, ferric sesquisulfateShow MoreShow Less
PubChem CID 24826
ChEBI CHEBI:53438
IUPAC Name iron(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3]

Specifications

Melting Point 480°C (decomposition)
Physical Form Powder
Quantity 5 g
Merck Index 14,4032
Solubility Information Soluble in water; Sparingly soluble in alcohol; Almost insoluble in acetone and ethyl acetate; Insoluble in sulfuric acid and ammonia.
Formula Weight 399.87
Percent Purity 99.998%
Grade Puratronic™
Odor Odorless
Sensitivity Hygroscopic
Density 3.097 g/mL
Assay (metals basis)
Chemical Name or Material Iron(III) sulfate
Show More Show Less
compliance-icons
Product Identifier
  • Iron(III) sulfate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(III) sulfate, Puratronic™, 99.998% (metals basis), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.