missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ISO 7010 Sign - Evacuation assembly point

Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.

Brand:  Brady™ 814927

Additional Details : Weight : 0.01670kg

 View more versions of this product

Product Code. 15604317

  • £19.85 / Each
Estimated Shipment: 15-08-2024
to see stock.
Add to basket

Description

Description

  • Polypropylene
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
Indoor and Outdoor
1.501 mm
-40°C to 120°C
Square
White on Green
Polypropylene
400 mm
400 mm
400 mm
Global
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers