missing translation for 'onlineSavingsMsg'
Learn More

Hopen™ Iso Air Cleanroom Masks with Headloops
GREENER_CHOICE

ISO Air cleanroom masks are ISO 3 Grade B and easy to put on with increased comfort thanks to its double elastic technology. Made with Mass Balance approach from recycled materials.

Brand:  Hopen™ 60.701-30

 View more versions of this product

Product Code. 17784779

  • £130.00 / Pack of 300
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Low particle release: Helmke drum tested for use in clean rooms
  • High filtration efficiency of particles, bacteria and viruses
  • Adaptable to users and protocols with two sizes available
  • Headloops improve comfort for the user by making it easier to put on
  • Softex technology limits risk of particulate contamination while reducing risk of skin irritation for the user
  • Origami folding and double elastics technology ensure optimal fit
  • Adjustable and malleable nose clip prevents glasses from fogging up
  • ISO AIR range is manufactured in an ISO 8 classified clean room
  • Made in France
Specifications

Specifications

Cleanroom Face Mask
Headband
230 x 90 mm
Grade B
>99%
60.701
White
Large
Pleated
Class 1 (ISO 3)
Industry
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers