missing translation for 'onlineSavingsMsg'
Learn More

Hopen™ Iso Air Duckbill Cleanroom Masks
GREENER_CHOICE

ISO Air duckbill cleanroom masks provide a large breathing chamber and are made with the Mass Balance approach from recycled materials.

Brand:  Hopen™ 60.754M

 View more versions of this product

Product Code. 17754789

  • £140.00 / Pack of 300
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Low particle release: Helmke drum tested for use in clean rooms
  • High filtration efficiency of particles, bacteria and viruses
  • Adaptable to users and protocols with two sizes available
  • Highly breathable thanks to a large breathing chamber
  • Softex technology limits risk of particulate contamination while reducing risk of skin irritation for the user
  • Adjustable and malleable nose clip prevents glasses from fogging up
  • ISO AIR range is manufactured in an ISO 8 classified clean room
  • Made in France
Specifications

Specifications

Cleanroom Face Mask
Medium
Grade B
Industry
White
Pouch
Class 1 (ISO 3)
60.754
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers